siRNA Details
| Gene Symbol: INTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020748.2 | NP_065799.1 | 16, 17 | 2195 | ||
| NR_026641.1 | 16, 17 | 2476 | ||||
| XM_006721987.3 | XP_006722050.1 | 16, 17 | 2202 | |||
| XM_011525058.2 | XP_011523360.1 | 16, 17 | 2482 | |||
| GenBank mRNA | AB033113.1 | - | 16, 17 | 2423 | ||
| AK299032.1 | - | 16, 17 | 2451 | |||
| AK308307.1 | - | 16, 17 | 2160 | |||
| AL136800.1 | - | 16, 17 | 2195 | |||
| CR533582.1 | - | 15, 16 | 2119 | |||

INTS2
Genomic Map