siRNA Details
| Gene Symbol: COG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020751.2 | NP_065802.1 | 18, 19 | 1927 | ||
| NR_026745.1 | 19, 20 | 1992 | ||||
| GenBank mRNA | AF116827.1 | - | 19, 20 | 1875 | ||
| AK026638.1 | - | 17, 18 | 1594 | |||
| AK294443.1 | - | 18, 19 | 1927 | |||
| BC051723.1 | - | 18, 19 | 1877 | 5263056 | ||
| CR627406.1 | - | 18, 19 | 1870 | |||

COG6
Genomic Map