siRNA Details
| Gene Symbol: COG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145079.1 | NP_001138551.1 | 5 | 558 | ||
| NM_020751.2 | NP_065802.1 | 5 | 558 | |||
| NR_026745.1 | 6 | 623 | ||||
| XM_011535168.1 | XP_011533470.1 | 5 | 508 | |||
| GenBank mRNA | AB032960.1 | - | 5 | 448 | ||
| AF116827.1 | - | 6 | 506 | |||
| AK026638.1 | - | 4 | 225 | |||
| AK294443.1 | - | 5 | 558 | |||
| BC051723.1 | - | 5 | 508 | 5263056 | ||
| CR627406.1 | - | 5 | 501 | |||

COG6
Genomic Map