siRNA Details
| Gene Symbol: KLHL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020782.1 | NP_065833.1 | 1 | 464 | ||
| XM_017019698.1 | XP_016875187.1 | 1 | 761 | |||
| GenBank mRNA | AB037761.1 | - | 1 | 195 | ||
| AK095405.1 | - | 1 | 464 | |||
| BC028742.1 | - | 2 | 332 | |||
| BC108669.1 | - | 1 | 435 | 6173072 | ||
| BC137116.1 | - | 1 | 430 | 9021113 | ||
| BC137117.1 | - | 1 | 430 | 9021114 | ||

KLHL42
Genomic Map