siRNA Details
| Gene Symbol: CEP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020802.3 | NP_065853.3 | 5 | 892 | ||
| XM_005271627.4 | XP_005271684.2 | 5 | 623 | |||
| GenBank mRNA | AB037798.1 | - | 2 | 235 | ||
| AK095004.1 | - | 4 | 522 | |||
| AK296350.1 | - | 2 | 633 | |||
| AK310418.1 | - | 4 | 1416 | |||
| BX647415.1 | - | 5 | 642 | |||

CEP126
Genomic Map