siRNA Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020825.3 | NP_065876.3 | 19, 20 | 3641 | ||
| GenBank mRNA | AB037847.2 | - | 17, 18 | 3034 | ||
| BC137172.1 | - | 18, 19 | 3191 | 9021169 | ||
| BC137173.1 | - | 18, 19 | 3191 | 9021170 | ||
| BC152477.1 | - | 17, 18 | 3034 | 8862615 | ||

CRAMP1
Genomic Map