siRNA Details
| Gene Symbol: RANBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320239.1 | NP_001307168.1 | 2, 3 | 467 | ||
| NM_001320240.1 | NP_001307169.1 | 2, 3 | 467 | |||
| NM_020850.2 | NP_065901.1 | 2, 3 | 467 | |||
| XM_005256070.2 | XP_005256127.1 | 2, 3 | 375 | |||
| XM_017023508.1 | XP_016878997.1 | 2, 3 | 375 | |||
| GenBank mRNA | AB040897.1 | - | 2, 3 | 356 | ||
| AK055727.1 | - | 2, 3 | 372 | |||
| AK298806.1 | - | 2, 3 | 394 | |||
| AK304030.1 | - | 2, 3 | 372 | |||
| AY337313.1 | - | 2, 3 | 467 | |||
| BC099917.1 | - | 2, 3 | 372 | 30528481 | ||
| BC121176.1 | - | 2, 3 | 465 | 40122561 | ||
| BC121177.2 | - | 2, 3 | 465 | 40122562 | ||

RANBP10
Genomic Map