siRNA Details
| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286725.1 | NP_001273654.1 | 3 | 1582 | ||
| NM_020866.2 | NP_065917.1 | 4 | 1765 | |||
| XM_017020678.1 | XP_016876167.1 | 4 | 685 | |||
| XM_017020679.1 | XP_016876168.1 | 4 | 1272 | |||
| GenBank mRNA | AB040923.1 | - | 4 | 1731 | ||
| AF252283.1 | - | 4 | 1599 | |||
| AK054609.1 | - | 4 | 977 | |||
| AK291435.1 | - | 4 | 1765 | |||
| AK295939.1 | - | 3 | 773 | |||
| BC022460.1 | - | 4 | 1009 | 4819077 | ||

KLHL1
Genomic Map