siRNA Details
| Gene Symbol: EP400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015409.4 | NP_056224.3 | 38 | 7047 | ||
| GenBank mRNA | AB058721.1 | - | 9 | 994 | ||
| AK074395.1 | - | 10 | 1240 | |||
| AY044869.1 | - | 37 | 6931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EP400| Gene Symbol: EP400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015409.4 | NP_056224.3 | 38 | 7047 | ||
| GenBank mRNA | AB058721.1 | - | 9 | 994 | ||
| AK074395.1 | - | 10 | 1240 | |||
| AY044869.1 | - | 37 | 6931 | |||