siRNA Details
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 64 | 15122 | ||
XM_005257545.4 | XP_005257602.2 | 65 | 15310 | |||
XM_005257546.4 | XP_005257603.2 | 65 | 15214 | |||
XM_006721995.3 | XP_006722058.1 | 65 | 15597 | |||
XM_017024905.1 | XP_016880394.1 | 60 | 14177 | |||
GenBank mRNA | AB046774.1 | - | 24 | 3312 | ||
AB537889.1 | - | 64 | 15115 | |||
AK023871.1 | - | 7 | 941 | |||
AK025676.1 | - | 18 | 2528 | |||
AK025914.1 | - | 17 | 2296 | |||
AK026038.1 | - | 3 | 308 | |||
AK074030.1 | - | 26 | 3666 | |||
AL161961.1 | - | 3 | 297 | |||
AL833201.1 | - | 24 | 3340 | |||
BC032220.1 | - | 8 | 1086 | |||
BX640932.1 | - | 30 | 4278 | |||
BX647946.1 | - | 36 | 5913 |
