siRNA Details
| Gene Symbol: VAT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020927.2 | NP_065978.1 | 2 | 426 | ||
| GenBank mRNA | AB046796.1 | - | 2 | 396 | ||
| AK290153.1 | - | 2 | 404 | |||
| AL834240.1 | - | 1 | 41 | |||
| BC033537.1 | - | 2 | 404 | 4821265 | ||
| HY132852.1 | - | 2 | 427 | |||

VAT1L
Genomic Map