siRNA Details
| Gene Symbol: GBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020944.2 | NP_065995.1 | 3 | 1037 | ||
| XM_005251526.4 | XP_005251583.1 | 3 | 1064 | |||
| XM_006716809.3 | XP_006716872.1 | 3 | 1064 | |||
| XM_011517973.2 | XP_011516275.1 | 3 | 642 | |||
| XM_017014937.1 | XP_016870426.1 | 3 | 1064 | |||
| XM_017014938.1 | XP_016870427.1 | 3 | 1064 | |||
| XM_017014939.1 | XP_016870428.1 | 3 | 642 | |||
| XM_017014940.1 | XP_016870429.1 | 3 | 642 | |||
| XM_017014941.1 | XP_016870430.1 | 3 | 1064 | |||
| XM_017014942.1 | XP_016870431.1 | 3 | 642 | |||
| XM_017014943.1 | XP_016870432.1 | 3 | 642 | |||
| XM_017014944.1 | XP_016870433.1 | 1 | 36 | |||
| XM_017014945.1 | XP_016870434.1 | 1 | 36 | |||
| XM_017014946.1 | XP_016870435.1 | 3 | 642 | |||
| GenBank mRNA | AB046825.1 | - | 3 | 1027 | ||
| AJ309567.1 | - | 3 | 1037 | |||
| AK027884.1 | - | 3 | 700 | |||
| AK295967.1 | - | 4 | 1012 | |||
| AK297435.1 | - | 1 | 26 | |||
| AL834306.1 | - | 3 | 797 | |||
| BC011363.1 | - | 3 | 996 | 4339156 | ||

GBA2
Genomic Map