siRNA Details
| Gene Symbol: SLC7A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020949.2 | NP_066000.2 | 3 | 778 | ||
| GenBank mRNA | AB046833.1 | - | 2 | 229 | ||
| AK122655.1 | - | 3 | 773 | |||
| BC022968.1 | - | 2 | 394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC7A14| Gene Symbol: SLC7A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020949.2 | NP_066000.2 | 3 | 778 | ||
| GenBank mRNA | AB046833.1 | - | 2 | 229 | ||
| AK122655.1 | - | 3 | 773 | |||
| BC022968.1 | - | 2 | 394 | |||