siRNA Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020961.3 | NP_066012.1 | 8 | 842 | ||
| XM_005263166.4 | XP_005263223.1 | 8 | 837 | |||
| XM_011532162.2 | XP_011530464.1 | 8 | 837 | |||
| GenBank mRNA | AB046847.1 | - | 8 | 797 | ||
| AK055555.1 | - | 8 | 820 | |||
| AK296056.1 | - | 6 | 462 | |||
| BC006565.2 | - | 8 | 793 | 3010654 | ||
| BC007449.2 | - | 8 | 793 | 3010654 | ||

METTL14
Genomic Map