siRNA Details
| Gene Symbol: BRINP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021165.3 | NP_066988.1 | 8 | 1669 | ||
| XM_005245379.1 | XP_005245436.1 | 9 | 2249 | |||
| XM_011509826.1 | XP_011508128.1 | 6 | 1496 | |||
| GenBank mRNA | AB051534.1 | - | 8 | 1662 | ||
| AB161694.1 | - | 8 | 1662 | |||
| AK123336.1 | - | 5 | 1104 | |||
| AK296524.1 | - | 9 | 1745 | |||
| AL035289.1 | - | 7 | 1243 | |||
| BC028036.1 | - | 8 | 1561 | 5242899 | ||

BRINP2
Genomic Map