siRNA Details
| Gene Symbol: ZP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021186.4 | NP_067009.1 | 9 | 1462 | ||
| GenBank mRNA | AK314151.1 | - | 9 | 1462 | ||
| BC069521.1 | - | 9 | 1175 | 7262334 | ||
| BE439656.1 | - | 4 | 494 | |||
| DV080723.1 | - | 2 | 125 | |||

ZP4
Genomic Map