siRNA Details
| Gene Symbol: NEUROD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021191.2 | NP_067014.2 | 2 | 1228 | ||
| GenBank mRNA | AK314136.1 | - | 2 | 1143 | ||
| BC040961.1 | - | 1 | 718 | 5490979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEUROD4| Gene Symbol: NEUROD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021191.2 | NP_067014.2 | 2 | 1228 | ||
| GenBank mRNA | AK314136.1 | - | 2 | 1143 | ||
| BC040961.1 | - | 1 | 718 | 5490979 | ||