siRNA Details
| Gene Symbol: MS4A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021201.4 | NP_067024.1 | 7 | 902 | ||
| NM_206938.1 | NP_996821.1 | 6 | 767 | |||
| NM_206939.1 | NP_996822.1 | 7 | 854 | |||
| NM_206940.1 | NP_996823.1 | 6 | 719 | |||
| GenBank mRNA | AB026043.1 | - | 7 | 842 | ||
| AF201951.1 | - | 7 | 813 | |||
| AF237916.1 | - | 7 | 826 | |||
| AF309653.1 | - | 7 | 855 | |||
| AF350501.1 | - | 7 | 813 | |||
| AK075106.1 | - | 7 | 906 | |||
| AK297027.1 | - | 6 | 703 | |||
| BC020673.1 | - | 7 | 826 | 4720044 | ||

MS4A7
Genomic Map