siRNA Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292017.1 | NP_001278946.1 | 4 | 826 | ||
| NM_021204.4 | NP_067027.1 | 4 | 855 | |||
| NR_120457.1 | 3 | 623 | ||||
| GenBank mRNA | AF087881.1 | - | 4 | 725 | ||
| AF113125.1 | - | 4 | 726 | |||
| AF177286.1 | - | 2 | 292 | |||
| AK022656.1 | - | 4 | 721 | |||
| AL518796.3 | - | 4 | 658 | |||
| BC001317.1 | - | 3 | 441 | |||
| BC065815.1 | - | 4 | 693 | 6526329 | ||
| CR457141.1 | - | 4 | 505 | |||

ENOPH1
Genomic Map