siRNA Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303461.1 | NP_001290390.1 | 4 | 840 | ||
| NM_001303462.1 | NP_001290391.1 | 3 | 429 | |||
| NM_021220.3 | NP_067043.2 | 4 | 889 | |||
| GenBank mRNA | AK022284.1 | - | 4 | 884 | ||
| AL079276.1 | - | 3 | 469 | |||
| BC006148.1 | - | 4 | 889 | 3959436 | ||
| BM563561.1 | - | 3 | 429 | |||
| BT007295.1 | - | 4 | 641 | |||
| BX331154.2 | - | 1 | 552 | |||

OVOL2
Genomic Map