siRNA Details
| Gene Symbol: ZNF250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109689.3 | NP_001103159.1 | 6 | 617 | ||
| NM_021061.4 | NP_066405.1 | 6 | 632 | |||
| XM_005272327.1 | XP_005272384.1 | 6 | 644 | |||
| XM_005272328.3 | XP_005272385.1 | 6 | 1117 | |||
| XM_006716612.3 | XP_006716675.1 | 6 | 1132 | |||
| XM_006716613.3 | XP_006716676.1 | 6 | 671 | |||
| XM_011517209.1 | XP_011515511.1 | 6 | 644 | |||
| GenBank mRNA | AB208810.1 | - | 6 | 587 | ||
| AK054888.1 | - | 6 | 632 | |||
| AK095705.1 | - | 6 | 575 | |||
| BC017091.1 | - | 6 | 614 | 3851128 | ||

ZNF250
Genomic Map