siRNA Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302807.1 | NP_001289736.1 | 4 | 489 | ||
| NM_021225.4 | NP_067048.4 | 3 | 433 | |||
| NR_126503.1 | 2 | 367 | ||||
| GenBank mRNA | CD722615.1 | - | 3 | 433 | ||
| CD723077.1 | - | 4 | 416 | |||
| CK429697.1 | - | 2 | 284 | |||
| S83198.1 | - | 3 | 339 | |||

OPRPN
Genomic Map