siRNA Details
| Gene Symbol: RRAGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021244.4 | NP_067067.1 | 7 | 1364 | ||
| XM_005248755.4 | XP_005248812.1 | 6 | 1409 | |||
| XM_011536022.2 | XP_011534324.1 | 7 | 1136 | |||
| GenBank mRNA | AF272036.1 | - | 7 | 1147 | ||
| AK126164.1 | - | 1 | 2440 | |||
| AK289799.1 | - | 7 | 1289 | |||
| AL137502.1 | - | 6 | 862 | |||
| BC003088.1 | - | 7 | 1026 | 3536088 | ||

RRAGD
Genomic Map