siRNA Details
| Gene Symbol: HRH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143828.1 | NP_001137300.1 | 2 | 710 | ||
| NM_001160166.1 | NP_001153638.1 | 2 | 810 | |||
| NM_021624.3 | NP_067637.2 | 3 | 974 | |||
| GenBank mRNA | AB044934.1 | - | 3 | 928 | ||
| AB045370.1 | - | 3 | 937 | |||
| AF312230.1 | - | 3 | 974 | |||
| AF325356.1 | - | 3 | 874 | |||
| AF329449.1 | - | 3 | 874 | |||
| AJ298292.1 | - | 3 | 874 | |||
| AY008280.1 | - | 3 | 874 | |||
| AY136745.1 | - | 3 | 874 | |||
| BC069136.1 | - | 3 | 974 | 7216992 | ||
| BC112348.1 | - | 3 | 934 | 40006540 | ||
| DQ835186.1 | - | 2 | 610 | |||
More Information
Set Membership: |
|

HRH4
Genomic Map