siRNA Details
| Gene Symbol: LGR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017403.1 | NP_001017403.1 | 18 | 2638 | ||
| NM_001017404.1 | NP_001017404.1 | 16 | 2221 | |||
| NM_021636.2 | NP_067649.2 | 18 | 2525 | |||
| XM_005245404.3 | XP_005245461.1 | 18 | 2582 | |||
| XM_011509838.2 | XP_011508140.1 | 17 | 2511 | |||
| XM_011509839.2 | XP_011508141.1 | 17 | 2512 | |||
| XM_011509840.2 | XP_011508142.1 | 17 | 2367 | |||
| XM_011509841.2 | XP_011508143.1 | 15 | 2262 | |||
| XM_011509842.2 | XP_011508144.1 | 15 | 3990 | |||
| XM_011509843.2 | XP_011508145.1 | 15 | 2382 | |||
| XM_011509844.2 | XP_011508146.1 | 12 | 1891 | |||
| XM_011509846.2 | XP_011508148.1 | 15 | 2287 | |||
| XM_017001996.1 | XP_016857485.1 | 16 | 2440 | |||
| XM_017001997.1 | XP_016857486.1 | 16 | 2397 | |||
| XM_017001998.1 | XP_016857487.1 | 7 | 1737 | |||
| GenBank mRNA | AB049405.1 | - | 18 | 2523 | ||
| AF190501.1 | - | 16 | 2221 | |||
| AK027377.1 | - | 1 | 933 | |||
| AK123055.1 | - | 12 | 1076 | |||
| AY358119.1 | - | 18 | 2566 | |||
| BC047905.1 | - | 18 | 2508 | 5212508 | ||

LGR6
Genomic Map