siRNA Details
| Gene Symbol: C12orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021640.3 | NP_067653.3 | 6 | 830 | ||
| GenBank mRNA | AA972697.1 | - | 2 | 384 | ||
| AF111805.1 | - | 4 | 422 | |||
| AF289485.1 | - | 6 | 798 | |||
| BC013956.2 | - | 5 | 617 | |||
| BC028904.1 | - | 5 | 734 | 4454943 | ||
| BC051871.1 | - | 6 | 789 | 5170816 | ||
| BM551107.1 | - | 2 | 136 | |||

C12orf10
Genomic Map