siRNA Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021733.1 | NP_068379.1 | 10 | 1606 | ||
| XM_011527201.1 | XP_011525503.1 | 10 | 1719 | |||
| XM_011527202.1 | XP_011525504.1 | 10 | 1719 | |||
| XM_011527203.2 | XP_011525505.1 | 10 | 1676 | |||
| GenBank mRNA | AF411384.1 | - | 6 | 1145 | ||
| BC058862.1 | - | 10 | 1634 | 5170446 | ||

TSKS
Genomic Map