siRNA Details
| Gene Symbol: SAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021818.3 | NP_068590.1 | 3 | 1116 | ||
| XM_011537057.2 | XP_011535359.1 | 4 | 1179 | |||
| XM_017021568.1 | XP_016877057.1 | 4 | 1409 | |||
| GenBank mRNA | AJ292969.1 | - | 3 | 895 | ||
| AK021500.1 | - | 3 | 1063 | |||
| AK023071.1 | - | 3 | 945 | |||
| AK095903.1 | - | 4 | 980 | |||
| AK225896.1 | - | 3 | 1088 | |||
| AK290883.1 | - | 4 | 988 | |||
| AL833378.1 | - | 2 | 526 | |||
| BC020537.1 | - | 3 | 1062 | 4425620 | ||
| CR457297.1 | - | 3 | 730 | |||

SAV1
Genomic Map