siRNA Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021928.3 | NP_068747.1 | 4 | 451 | ||
| GenBank mRNA | AK026302.1 | - | 4 | 426 | ||
| AK092634.1 | - | 3 | 1434 | |||
| AL136660.1 | - | 4 | 421 | |||
| AY359044.1 | - | 4 | 409 | |||
| BC047290.1 | - | 4 | 394 | 5224927 | ||
| BC053883.1 | - | 3 | 2345 | |||
| DB285025.1 | - | 4 | 453 | |||

SPCS3
Genomic Map