siRNA Details
| Gene Symbol: RIC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286134.1 | NP_001273063.1 | 3 | 1631 | ||
| NM_021932.5 | NP_068751.4 | 3 | 1631 | |||
| XM_005253052.3 | XP_005253109.1 | 3 | 817 | |||
| XM_017018107.1 | XP_016873596.1 | 3 | 901 | |||
| GenBank mRNA | AK022870.1 | - | 2 | 185 | ||
| AK026897.1 | - | 3 | 1630 | |||
| AK027461.1 | - | 3 | 319 | |||
| AK225420.1 | - | 3 | 320 | |||
| AK314568.1 | - | 3 | 341 | |||
| AL136935.1 | - | 3 | 612 | |||
| AL390088.1 | - | 3 | 352 | |||
| BC011821.1 | - | 1 | 98 | |||
| BC111499.1 | - | 3 | 448 | 6526180 | ||
| BC121807.1 | - | 3 | 528 | 40108083 | ||
| BC121808.1 | - | 3 | 528 | 40108080 | ||
| BX538115.1 | - | 1 | 493 | |||

RIC8A
Genomic Map