siRNA Details
| Gene Symbol: ATG101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098673.1 | NP_001092143.1 | 4 | 824 | ||
| NM_021934.4 | NP_068753.2 | 4 | 995 | |||
| GenBank mRNA | AF218031.1 | - | 4 | 935 | ||
| AK021835.1 | - | 4 | 993 | |||
| BC005151.2 | - | 4 | 947 | 3508173 | ||
| BC009937.2 | - | 4 | 929 | 4303452 | ||

ATG101
Genomic Map