siRNA Details
| Gene Symbol: CELF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021938.3 | NP_068757.2 | 10, 11 | 1234 | ||
| XM_006722832.1 | XP_006722895.1 | 10, 11 | 1261 | |||
| XM_006722833.1 | XP_006722896.1 | 10, 11 | 1258 | |||
| XM_006722836.2 | XP_006722899.1 | 9, 10 | 902 | |||
| XM_006722837.3 | XP_006722900.1 | 10, 11 | 1173 | |||
| XM_006722838.3 | XP_006722901.1 | 9, 10 | 1045 | |||
| XM_011528173.1 | XP_011526475.1 | 9, 10 | 874 | |||
| XM_011528174.2 | XP_011526476.1 | 9, 10 | 1043 | |||
| XM_017027111.1 | XP_016882600.1 | 8, 9 | 1117 | |||
| GenBank mRNA | AF248649.1 | - | 1 | 13 | ||
| AF329266.1 | - | 10, 11 | 1197 | |||
| BC028101.1 | - | 10, 11 | 1234 | 5242909 | ||

CELF5
Genomic Map