siRNA Details
| Gene Symbol: HEATR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022070.4 | NP_071353.4 | 18 | 2725 | ||
| XM_005257570.3 | XP_005257627.1 | 19 | 2913 | |||
| XM_011525110.2 | XP_011523412.1 | 17 | 2612 | |||
| XM_011525111.2 | XP_011523413.1 | 17 | 2574 | |||
| XM_017024936.1 | XP_016880425.1 | 18 | 2986 | |||
| XM_017024937.1 | XP_016880426.1 | 18 | 4662 | |||
| XM_017024938.1 | XP_016880427.1 | 18 | 2718 | |||
| XM_017024939.1 | XP_016880428.1 | 18 | 2989 | |||
| GenBank mRNA | AF349752.1 | - | 19 | 2670 | ||
| AK025740.1 | - | 11 | 1792 | |||
| AK026070.1 | - | 11 | 1565 | |||
| AK127708.1 | - | 18 | 2719 | |||
| AK314655.1 | - | 18 | 2728 | |||
| BC119771.1 | - | 16 | 2379 | 40108435 | ||
| BC119813.1 | - | 18 | 2716 | 40108426 | ||
| BX640819.1 | - | 18 | 2716 | |||
| CR627418.1 | - | 18 | 2715 | |||

HEATR6
Genomic Map