siRNA Details
| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022095.3 | NP_071378.1 | 17 | 2511 | ||
| XM_005260504.4 | XP_005260561.1 | 17 | 3224 | |||
| XM_011528979.2 | XP_011527281.1 | 16 | 2879 | |||
| XM_017028012.1 | XP_016883501.1 | 16 | 2282 | |||
| GenBank mRNA | AF395833.1 | - | 17 | 2488 | ||
| AK026157.1 | - | 9 | 1026 | |||
| AK074214.1 | - | 2 | 146 | |||
| AK296989.1 | - | 14 | 2082 | |||
| BC041331.2 | - | 14 | 1726 | |||
| BC142981.1 | - | 14 | 1726 | |||

ZNF335
Genomic Map