siRNA Details
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 2 | 376 | ||
NM_001170570.1 | NP_001164041.1 | 2 | 294 | |||
NM_022101.3 | NP_071384.1 | 2 | 294 | |||
GenBank mRNA | AK096297.1 | - | 2 | 340 | ||
AK298877.1 | - | 2 | 272 | |||
BC023506.2 | - | 2 | 255 | |||
BQ223940.1 | - | 2 | 259 | |||
DA590933.1 | - | 2 | 378 | |||
DA921725.1 | - | 2 | 269 | |||
DB033119.1 | - | 2 | 309 |
