siRNA Details
| Gene Symbol: DMRTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033067.2 | NP_149056.1 | 1 | 211 | ||
| GenBank mRNA | AJ291671.1 | - | 1 | 138 | ||
| AK057273.1 | - | 1 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMRTB1| Gene Symbol: DMRTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033067.2 | NP_149056.1 | 1 | 211 | ||
| GenBank mRNA | AJ291671.1 | - | 1 | 138 | ||
| AK057273.1 | - | 1 | 211 | |||