siRNA Details
| Gene Symbol: LOC107986113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007620.1 | XP_016863109.1 | 2 | 1359 | ||
| Gene Symbol: ERVK-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | JZ164911.1 | - | 1 | 528 | ||

LOC107986113
Genomic Map