siRNA Details
| Gene Symbol: MMP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022122.2 | NP_071405.2 | 5 | 771 | ||
| XM_011542948.2 | XP_011541250.1 | 4 | 690 | |||
| XM_011542950.2 | XP_011541252.1 | 4 | 517 | |||
| XM_017018120.1 | XP_016873609.1 | 5 | 819 | |||
| GenBank mRNA | AF195192.1 | - | 5 | 754 | ||
| AY358752.1 | - | 5 | 771 | |||
| BX102293.1 | - | 1 | 19 | |||

MMP27
Genomic Map