siRNA Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022144.2 | NP_071427.2 | 2, 3 | 417 | ||
| GenBank mRNA | AB055421.1 | - | 2, 3 | 197 | ||
| AF191770.1 | - | 2, 3 | 377 | |||
| AF234259.1 | - | 2, 3 | 258 | |||
| AF291656.1 | - | 2, 3 | 263 | |||
| AY358706.1 | - | 2, 3 | 249 | |||
| BP319044.1 | - | 2, 3 | 417 | |||

TNMD
Genomic Map