siRNA Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022158.3 | NP_071441.1 | 1 | 180 | ||
| GenBank mRNA | AJ404615.1 | - | 1 | 140 | ||
| AK294871.1 | - | 1 | 141 | |||
| BC042680.1 | - | 1 | 124 | 5195472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FN3K| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022158.3 | NP_071441.1 | 1 | 180 | ||
| GenBank mRNA | AJ404615.1 | - | 1 | 140 | ||
| AK294871.1 | - | 1 | 141 | |||
| BC042680.1 | - | 1 | 124 | 5195472 | ||