siRNA Details
| Gene Symbol: C17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022344.3 | NP_071739.2 | 6 | 691 | ||
| XM_005258022.3 | XP_005258079.1 | 7 | 709 | |||
| XM_017024940.1 | XP_016880429.1 | 6 | 560 | |||
| XM_017024941.1 | XP_016880430.1 | 6 | 558 | |||
| GenBank mRNA | AF052093.1 | - | 6 | 618 | ||
| AF305686.1 | - | 6 | 647 | |||
| BC026017.2 | - | 6 | 637 | 3352761 | ||
| BC035715.1 | - | 6 | 645 | 5588164 | ||
| BC047325.1 | - | 6 | 631 | 5493477 | ||
| BC054035.1 | - | 6 | 625 | 6143106 | ||

C17orf75
Genomic Map