siRNA Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129758.1 | NP_001123230.1 | 5, 6 | 930 | ||
| NM_022357.3 | NP_071752.3 | 5, 6 | 930 | |||
| GenBank mRNA | AJ291679.1 | - | 5, 6 | 844 | ||
| AK057401.1 | - | 5, 6 | 930 | |||
| AY358390.1 | - | 5, 6 | 853 | |||
| BC037243.1 | - | 5, 6 | 842 | |||
| BC057789.1 | - | 5, 6 | 871 | 5297958 | ||

DPEP3
Genomic Map