siRNA Details
| Gene Symbol: DNAJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022365.3 | NP_071760.2 | 11 | 1507 | ||
| GenBank mRNA | AF427142.1 | - | 4 | 383 | ||
| AK026062.1 | - | 8 | 780 | |||
| AK027263.1 | - | 11 | 1327 | |||
| AY225122.1 | - | 11 | 1220 | |||
| BC030955.1 | - | 7 | 690 | |||
| BC110894.1 | - | 11 | 1507 | 5164559 | ||
| BC139752.1 | - | 3 | 227 | |||

DNAJC1
Genomic Map