siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 17 | 2174 | ||
| XM_011545916.2 | XP_011544218.1 | 16 | 3521 | |||
| XM_011545917.2 | XP_011544219.1 | 16 | 3521 | |||
| XM_011545919.2 | XP_011544221.1 | 15 | 3484 | |||
| GenBank mRNA | AK021680.1 | - | 5 | 365 | ||
| AK022551.1 | - | 8 | 579 | |||
| AK093045.1 | - | 1 | 850 | |||
| AK096322.1 | - | 9 | 957 | |||
| AK298229.1 | - | 10 | 737 | |||
| BQ276414.1 | - | 10 | 742 | |||
| DR761499.1 | - | 10 | 671 | |||

FBRS
Genomic Map