siRNA Details
| Gene Symbol: RNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022453.2 | NP_071898.2 | 10 | 1060 | ||
| XM_017004695.1 | XP_016860184.1 | 10 | 1060 | |||
| GenBank mRNA | AK023968.1 | - | 10 | 978 | ||
| AK222525.1 | - | 10 | 998 | |||
| AK289501.1 | - | 10 | 1004 | |||
| BC015612.2 | - | 10 | 977 | 4811222 | ||

RNF25
Genomic Map