siRNA Details
| Gene Symbol: SIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037633.1 | NP_001032722.1 | 7 | 826 | ||
| NM_022464.4 | NP_071909.1 | 6 | 738 | |||
| XM_011543570.2 | XP_011541872.1 | 8 | 863 | |||
| GenBank mRNA | AF547994.1 | - | 6 | 657 | ||
| AJ299442.1 | - | 6 | 716 | |||
| AK074624.1 | - | 6 | 730 | |||
| AK075177.1 | - | 6 | 667 | |||
| AY358950.1 | - | 6 | 684 | |||
| BC011568.1 | - | 6 | 706 | 4640182 | ||
| KU178743.1 | - | 5 | 620 | |||

SIL1
Genomic Map