siRNA Details
| Gene Symbol: IKZF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022465.3 | NP_071910.3 | 8 | 1638 | ||
| XM_005269086.3 | XP_005269143.1 | 10 | 1180 | |||
| XM_005269089.2 | XP_005269146.1 | 8 | 1282 | |||
| XM_005269090.3 | XP_005269147.1 | 9 | 1086 | |||
| XM_005269093.2 | XP_005269150.1 | 7 | 1557 | |||
| XM_011538664.1 | XP_011536966.1 | 10 | 1183 | |||
| XM_011538666.2 | XP_011536968.1 | 9 | 1031 | |||
| XM_011538669.2 | XP_011536971.1 | 8 | 981 | |||
| XM_017019805.1 | XP_016875294.1 | 12 | 1428 | |||
| XM_017019806.1 | XP_016875295.1 | 12 | 1671 | |||
| XM_017019807.1 | XP_016875296.1 | 12 | 1668 | |||
| XM_017019808.1 | XP_016875297.1 | 11 | 1353 | |||
| XM_017019809.1 | XP_016875298.1 | 12 | 1425 | |||
| XM_017019810.1 | XP_016875299.1 | 12 | 1523 | |||
| XM_017019811.1 | XP_016875300.1 | 11 | 1350 | |||
| XM_017019812.1 | XP_016875301.1 | 10 | 1426 | |||
| XM_017019813.1 | XP_016875302.1 | 10 | 1423 | |||
| XM_017019814.1 | XP_016875303.1 | 9 | 1332 | |||
| XM_017019815.1 | XP_016875304.1 | 9 | 1329 | |||
| XM_017019816.1 | XP_016875305.1 | 9 | 1089 | |||
| GenBank mRNA | AB058685.1 | - | 6 | 1182 | ||
| AF230809.1 | - | 7 | 890 | |||
| AK122880.1 | - | 7 | 1405 | |||
| AK314543.1 | - | 7 | 1003 | |||
| BX647761.1 | - | 9 | 1208 | |||

IKZF4
Genomic Map