siRNA Details
| Gene Symbol: AKTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012398.2 | NP_001012398.1 | 6 | 738 | ||
| NM_001308325.1 | NP_001295254.1 | 6 | 709 | |||
| NM_022476.3 | NP_071921.1 | 6 | 709 | |||
| XM_005256095.4 | XP_005256152.1 | 6 | 1058 | |||
| XM_005256096.4 | XP_005256153.1 | 6 | 636 | |||
| XM_005256097.4 | XP_005256154.1 | 6 | 670 | |||
| XM_005256098.4 | XP_005256155.1 | 6 | 634 | |||
| XM_017023564.1 | XP_016879053.1 | 6 | 1058 | |||
| XM_017023565.1 | XP_016879054.1 | 6 | 636 | |||
| XM_017023566.1 | XP_016879055.1 | 6 | 634 | |||
| GenBank mRNA | AK023320.1 | - | 6 | 644 | ||
| AK222743.1 | - | 6 | 641 | |||
| BC001134.2 | - | 6 | 657 | 2967019 | ||
| BC095401.1 | - | 6 | 631 | 30334599 | ||
| CR457308.1 | - | 5 | 481 | |||

AKTIP
Genomic Map