siRNA Details
| Gene Symbol: TTC31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022492.4 | NP_071937.4 | 4 | 276 | ||
| NR_027749.1 | 3 | 173 | ||||
| XM_005264492.1 | XP_005264549.1 | 4 | 265 | |||
| XM_011533035.1 | XP_011531337.1 | 4 | 265 | |||
| XM_011533036.1 | XP_011531338.1 | 4 | 265 | |||
| XM_011533037.1 | XP_011531339.1 | 4 | 265 | |||
| XM_011533040.1 | XP_011531342.1 | 3 | 214 | |||
| XM_017004706.1 | XP_016860195.1 | 4 | 265 | |||
| XM_017004707.1 | XP_016860196.1 | 4 | 265 | |||
| XM_017004708.1 | XP_016860197.1 | 3 | 170 | |||
| XM_017004709.1 | XP_016860198.1 | 3 | 400 | |||
| GenBank mRNA | AK022850.1 | - | 4 | 262 | ||
| AK090520.1 | - | 3 | 315 | |||
| AK223355.1 | - | 4 | 260 | |||
| AK225626.1 | - | 4 | 262 | |||
| AK303105.1 | - | 3 | 159 | |||
| AK303245.1 | - | 4 | 262 | |||
| BC036017.1 | - | 4 | 260 | 5270575 | ||
| BC045536.1 | - | 3 | 164 | |||
| BC096709.1 | - | 4 | 262 | 40023059 | ||
| BC098365.1 | - | 4 | 262 | 40023060 | ||
| BC099729.1 | - | 3 | 159 | |||
| BX648131.1 | - | 3 | 158 | |||

TTC31
Genomic Map