siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033225.5 | NP_150094.5 | 28 | 4853 | ||
| XM_011534752.2 | XP_011533054.1 | 28 | 5082 | |||
| XM_011534753.2 | XP_011533055.1 | 11 | 1960 | |||
| XM_011534754.1 | XP_011533056.1 | 4 | 696 | |||
| XM_017013731.1 | XP_016869220.1 | 28 | 5082 | |||
| GenBank mRNA | AB067477.1 | - | 9 | 1571 | ||
| AB209502.1 | - | 16 | 2905 | |||
| AF333704.2 | - | 29 | 4856 | |||
| AK126936.1 | - | 4 | 696 | |||
| AY017307.1 | - | 29 | 4751 | |||
| AY358174.1 | - | 12 | 1996 | |||

CSMD1
Genomic Map